missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A4 Protein
A DNA sequence encoding the Homo sapiens (Human) S100A4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6639-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_002952.1 | |
S100A4 Protein | |
Research Use Only | |
S100A4 | |
Recombinant Protein | |
E. coli | |
ACPLEKALDV MVSTFHKYSG KEGDKFKLNK SELKELLTRE LPSFLGKRTD EAAFQKLMSN LDSNRDNEVD FQEYCVFLSC IAMMCNEFFE GFPDKQPRKK | |
Greater than 90% as determined by SDS-PAGE. |
6275 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |