missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Selenoprotein 1

A DNA sequence encoding the Homo sapiens (Human) Selenoprotein P, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6665-ec-100ug

 View more versions of this product

Product Code. 16024920

  • 6235.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001078955.1
Selenoprotein 1
Research Use Only
SEPP1
Recombinant Protein
E. coli
ESQDQSSLCK QPPAWSIRDQ DPMLNSNGSV TVVALLQASS YLCILQASKL EDLRVKLKKE GYSNISYIVV NHQGISSRLK YTHLKNKVSE HIPVYQQEEN QTDVWTLLNG SKDDFLIYDR CGRLVYHLGL PFSFLTFPYV EEAIKIAYCE KKCGNCSLTT LKDEDFCKRV SLATVDKTVE TPSPHYHHEH HHNHGHQHLG SSELSENQQP GAPNAPTHPA PPGLHHHHKH KGQHRQGHPE NRDMPASEDL QDLQKKLCRK RCINQLLCKL PTDSELAPRS SCCHCRHLIF EKTGSAITSQ CKENLPSLCS SQGLRAEENI TESCQSRLPP AASQISQQLI PTEASASSRS KNQAKKSESP SN
Greater than 90% as determined by SDS-PAGE.
6414
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers