missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal recognition particle 14 kDa Protein

A DNA sequence encoding the Homo sapiens (Human) Signal recognition particle 14 kDa protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8428-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024561

  • 5980.00 SEK / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_003125.3
Signal recognition particle 14 kDa Protein
Research Use Only
SRP14
Recombinant Protein
E. coli
VLLESEQFLT ELTRLFQKCR TSGSVYITLK KYDGRTKPIP KKGTVEGFEP ADNKCLLRAT DGKKKISTVV SSKEVNKFQM AYSNLLRANM DGLKKRDKKN KTKKTKA
Greater than 90% as determined by SDS-PAGE.
6727
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers