missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Signal recognition particle 14 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Signal recognition particle 14 kDa protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8428-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_003125.3 | |
Signal recognition particle 14 kDa Protein | |
Research Use Only | |
SRP14 | |
Recombinant Protein | |
E. coli | |
VLLESEQFLT ELTRLFQKCR TSGSVYITLK KYDGRTKPIP KKGTVEGFEP ADNKCLLRAT DGKKKISTVV SSKEVNKFQM AYSNLLRANM DGLKKRDKKN KTKKTKA | |
Greater than 90% as determined by SDS-PAGE. |
6727 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |