missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spindlin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Spindlin-1, was expressed in the hosts and tags indicated.

1580.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_006708.2
Gene ID (Entrez) 10927
Name Spindlin-1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16023821
View Documents Promotion Details
enQuireBio™
QP8292-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16063821
View Documents Promotion Details
enQuireBio™
QP8292-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16013821
View Documents Promotion Details
enQuireBio™
QP8292-ec-100ug
100 μg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16043821
View Documents Promotion Details
enQuireBio™
QP8292-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16053821
View Documents Promotion Details
enQuireBio™
QP8292-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16033821
View Documents Promotion Details
enQuireBio™
QP8292-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_006708.2
Spindlin-1 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
10927
Research Use Only
SPIN1
Recombinant Protein
E. coli
MKTPFGKTPG QRSRADAGHA GVSANMMKKR TSHKKHRSSV GPSKPVSQPR RNIVGCRIQH GWKEGNGPVT QWKGTVLDQV PVNPSLYLIK YDGFDCVYGL ELNKDERVSA LEVLPDRVAT SRISDAHLAD TMIGKAVEHM FETEDGSKDE WRGMVLARAP VMNTWFYITY EKDPVLYMYQ LLDDYKEGDL RIMPDSNDSP PAEREPGEVV DSLVGKQVEY AKEDGSKRTG MVIHQVEAKP SVYFIKFDDD FHIYVYDLVK TS
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications