missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Stathmin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Stathmin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8172-ec-50ug

 View more versions of this product

Product Code. 16083051

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001269982.1
Stathmin-4 Protein
Research Use Only
STMN4
Recombinant Protein
E. coli
MTLAAYKEKM KELPLVSLFC SCFLADPLNK SSYKYEGWCG RQCRRKDESQ RKDSADWRER RAQADTVDLN WCVISDMEVI ELNKCTSGQS FEVILKPPSF DGVPEFNASL PRRRDPSLEE IQKKLEAAEE RRKYQEAELL KHLAEKREHE REVIQKAIEE NNNFIKMAKE KLAQKMESNK ENREAHLAAM LERLQEKDKH AEEVRKNKEL KEEASR
Greater than 80% as determined by SDS-PAGE.
81551
50 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers