missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUB1 Protein
A DNA sequence encoding the Homo sapiens (Human) SUB1, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_006704.3 |
---|---|
Gene ID (Entrez) | 10923 |
Name | SUB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025530
|
enQuireBio™
QP6745-ec-10ug |
10 μg | N/A | ||||||
16065530
|
enQuireBio™
QP6745-ec-50ug |
50 μg | N/A | ||||||
16015530
|
enQuireBio™
QP6745-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16045530
|
enQuireBio™
QP6745-ec-200ug |
200 μg | N/A | ||||||
16055530
|
enQuireBio™
QP6745-ec-500ug |
500 μg | N/A | ||||||
16035530
|
enQuireBio™
QP6745-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_006704.3 | |
SUB1 Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
10923 | |
Research Use Only | |
SUB1 | |
Recombinant Protein | |
PKSKELVSSS SSGSDSDSEV DKKLKRKKQV APEKPVKKQK TGETSRALSS SKQSSSSRDD NMFQIGKMRY VSVRDFKGKV LIDIREYWMD PEGEMKPGRK GISLNPEQWS QLKEQISDID DAVRKL |