missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Surfactant-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Surfactant-associated protein 3, was expressed in the hosts and tags indicated.

1640.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_001094811.1
Gene ID (Entrez) 253970
Name Surfactant-associated protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16035040
View Documents
enQuireBio™
QP6680-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16075040
View Documents
enQuireBio™
QP6680-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16025040
View Documents
enQuireBio™
QP6680-ec-100ug
100 μg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16055040
View Documents
enQuireBio™
QP6680-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16065040
View Documents
enQuireBio™
QP6680-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16045040
View Documents
enQuireBio™
QP6680-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001094811.1
Surfactant-associated protein 1
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
253970
Research Use Only
SFTA3
Recombinant Protein
E. coli
MRAGFSDFQL IRDQVLFLQD QAQRLTEWLQ LSGFENPVSE STTLCLRERE KRIPTCVAVC VPSPGTVHTA LLHPTTLSQS RSSSEAKMLI IHTA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.