missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syntaxin-11 Protein

A DNA sequence encoding the Homo sapiens (Human) Syntaxin-11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6742-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075510

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_003755.2
Syntaxin-11 Protein
Research Use Only
STX11
Recombinant Protein
E. coli
MKDRLAELLD LSKQYDQQFP DGDDEFDSPH EDIVFETDHI LESLYRDIRD IQDENQLLVA DVKRLGKQNA RFLTSMRRLS SIKRDTNSIA KAIKARGEVI HCKLRAMKEL SEAAEAQHGP HSAVARISRA QYNALTLTFQ RAMHDYNQAE MKQRDNCKIR IQRQLEIMGK EVSGDQIEDM FEQGKWDVFS ENLLADVKGA RAALNEIESR HRELLRLESR IRDVHELFLQ MAVLVEKQAD TLNVIELNVQ KTVDYTGQAK AQVRKAVQYE EKNPCRTLCC FCCPCLK
Greater than 80% as determined by SDS-PAGE.
8676
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers