missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TALLA-1 / TSPAN7 Protein

A DNA sequence encoding the Homo sapiens (Human) TALLA-1 / TSPAN7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6846-ec-1mg

 View more versions of this product

Product Code. 16066250

  • 26990.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004606.2
TALLA-1 / TSPAN7 Protein
Research Use Only
TSPAN7
Recombinant Protein
E. coli
RHEIKDTFLR TYTDAMQTYN GNDERSRAVD HVQRSLSCCG VQNYTNWSTS PYFLEHGIPP SCCMNETDCN PQDLHNLTVA ATKVNQKGCY DLVTSFMETN M
Greater than 80.90% as determined by SDS-PAGE.
7102
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers