missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tetratricopeptide repeat protein 1

A DNA sequence encoding the Homo sapiens (Human) Tetratricopeptide repeat protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8348-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044151

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001269429.1
Tetratricopeptide repeat protein 1
Research Use Only
TTC1
Recombinant Protein
E. coli
EKSENCGVPE DLLNGLKVTD TQEAECAGPP VPDPKNQHSQ SKLLRDDEAH LQEDQGEEEC FHDCSASFEE EPGADKVENK SNEDVNSSEL DEEYLIELEK NMSDEEKQKR REESTRLKEE GNEQFKKGDY IEAESSYSRA LEMCPSCFQK ERSILFSNRA AARMKQDKKE MAINDCSKAI QLNPSYIRAI LRRAELYEKT DKLDEALEDY KSILEKDPSI HQAREACMRL PKQIEERNER LKEEMLGKLK DLGNLVLRPF GLSTENFQIK QDSSTGSYSI NFVQNPNNNR
Greater than 90% as determined by SDS-PAGE.
7265
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers