missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Titin Protein
A DNA sequence encoding the Homo sapiens (Human) Titin, was expressed in the hosts and tags indicated.
2185.00 SEK - 32135.00 SEK
Specifications
Accession Number | NP_001243779.1 |
---|---|
Gene ID (Entrez) | 7273 |
Name | Titin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026671
|
enQuireBio™
QP8823-EC-100UG |
100 μg |
7550.00 SEK
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046671
|
enQuireBio™
QP8823-EC-1MG |
1 mg |
32135.00 SEK
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036671
|
enQuireBio™
QP8823-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056671
|
enQuireBio™
QP8823-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066671
|
enQuireBio™
QP8823-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076671
|
enQuireBio™
QP8823-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001243779.1 | |
Titin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7273 | |
Research Use Only | |
TTN | |
Recombinant Protein | |
E. coli | |
VGSSICSARV TLREPPSFIK KIESTSSLRG GTAAFQATLK GSLPITVTWL KDSDEITEDD NIRMTFENNV ASLYLSGIEV KHDGKYVCQA KNDAGIQRCS ALLSVKEPAT ITEEAVSIDV TQGDPATLQV KFSGTKEITA KWFKDGQELT LGSKYKISVT DTVSILKIIS TEKKDSGEYT FEVQNDVGRS SCKARINVLD LIIPPSF | |
Greater than 90% as determined by SDS-PAGE. |