missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transmembrane 4 L6 family member 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transmembrane 4 L6 family member 1, was expressed in the hosts and tags indicated.
2955.00 SEK - 31480.00 SEK
Specifications
Accession Number | NP_055035.1 |
---|---|
Gene ID (Entrez) | 4071 |
Name | Transmembrane 4 L6 family member 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078301
|
enQuireBio™
QP9438-YE-100UG |
100 μg |
10410.00 SEK
100µg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018311
|
enQuireBio™
QP9438-YE-500UG |
500 μg |
29995.00 SEK
500µg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088301
|
enQuireBio™
QP9438-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008311
|
enQuireBio™
QP9438-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028311
|
enQuireBio™
QP9438-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098301
|
enQuireBio™
QP9438-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055035.1 | |
Transmembrane 4 L6 family member 1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4071 | |
Research Use Only | |
TM4SF1 | |
Recombinant Protein | |
Yeast | |
LAEGPLCLDS LGQWNYTFAS TEGQYLLDTS TWSECTEPKH IVEWNVS | |
Greater than 90% as determined by SDS-PAGE. |