missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trimethyllysine dioxygenase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Trimethyllysine dioxygenase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6808-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171726.1 | |
Trimethyllysine dioxygenase, mitochondrial Protein | |
Research Use Only | |
TMLHE | |
Recombinant Protein | |
E. coli | |
LLKGGVIYPA LPQPNFKSLL PLAVHWHHTA SKSLTCAWQQ HEDHFELKYA NTVMRFDYVW LRDHCRSASC YNSKTHQRSL DTASVDLCIK PKTIRLDETT LFFTWPDGHV TKYDLNWLVK NSYEGQKQKV IQPRILWNAE IYQQAQVPSV DCQSFLETNE GLKKFLQNFL LYGIAFVENV PPTQEHTEKL AERISLIRET IYGRMWYFTS DFSRGDTAYT KLALDRHTDT TYFQEPCGIQ VFHCLKHEGT GGRTLLVDGF YAAEQVLQKA PEEFELLSKV PLKHEYIEDV GECHNHMIGI GPVLNIYPWN KELYLIRLFK EKQNTVNRQW NSSLQCDIPE RILTYRHFVS GTSIEHRGSL I | |
Greater than 90% as determined by SDS-PAGE. |
55217 | |
100 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |