missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tumor protein D53 Protein
A DNA sequence encoding the Homo sapiens (Human) Tumor protein D53, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7623-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001003395.1 | |
Tumor protein D53 Protein | |
Research Use Only | |
TPD52L1 | |
Recombinant Protein | |
E. coli | |
MEAQAQGLLE TEPLQGTDED AVASADFSSM LSEEEKEELK AELVQLEDEI TTLRQVLSAK ERHLVEIKQK LGMNLMNELK QNFSKSWHDM QTTTAYKKTH ETLSHAGQKA TAAFSNVGTA ISKKFGDMSY SIRHSISMPA MRNSPTFKSF EERVETTVTS LKTKVGGTNP NGGSFEEVLS STAHASAQSL AGGSRRTKEE ELQC | |
Greater than 80% as determined by SDS-PAGE. |
7164 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |