missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tyrosine-protein kinase JAK1 Protein

A DNA sequence encoding the Homo sapiens (Human) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.

2850.00 SEK - 32610.00 SEK

Specifications

Accession Number NP_001307852.1
Gene ID (Entrez) 3716
Name Tyrosine-protein kinase JAK1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16081880
View Documents
enQuireBio™
QP6244-ye-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16021890
View Documents
enQuireBio™
QP6244-ye-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16071880
View Documents
enQuireBio™
QP6244-ye-100ug
100 μg
10850.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16001890
View Documents
enQuireBio™
QP6244-ye-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16011890
View Documents
enQuireBio™
QP6244-ye-500ug
500 μg
31245.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16091880
View Documents
enQuireBio™
QP6244-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_001307852.1
Tyrosine-protein kinase JAK1 Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
3716
Research Use Only
JAK1
Recombinant Protein
Yeast
EQNPDIVSEK KPATEVDPTH FEKRFLKRIR DLGEGHFGKV ELCRYDPEGD NTGEQVAVKS LKPESGGNHI ADLKKEIEIL RNLYHENIVK YKGICTEDGG NGIKLIMEFL PSGSLKEYLP KNKNKINLKQ QLKYAVQICK GMDYLGSRQY VHRDLAARNV LVESEHQVKI GDFGLTKAIE TDKEYYTVKD DRDSPVFWYA PECLMQSKFY IASDVWSFGV TLHELLTYCD SDSSPMALFL KMIGPTHGQM TVTRLVNTLK EGKRLPCPPN CPDEVYQLMR KCWEFQPSNR TSFQNLIEGF EALLK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.