missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Urokinase / PLAU Protein

A DNA sequence encoding the Homo sapiens (Human) Urokinase / PLAU, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8792-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026491

  • 5980.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001138503.1
Urokinase / PLAU Protein
Research Use Only
PLAU
Recombinant Protein
E. coli
SNELHQVPSN CDCLNGGTCV SNKYFSNIHW CNCPKKFGGQ HCEIDKSKTC YEGNGHFYRG KASTDTMGRP CLPWNSATVL QQTYHAHRSD ALQLGLGKHN YCRNPDNRRR PWCYVQVGLK PLVQECMVHD CADGKKPSSP PEELKFQCGQ KTL
Greater than 90% as determined by SDS-PAGE.
5328
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers