missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VWA5B2 Protein
A DNA sequence encoding the Homo sapiens (Human) VWA5B2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7823-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307302.1 | |
VWA5B2 Protein | |
Research Use Only | |
VWA5B2 | |
Recombinant Protein | |
E. coli | |
PRKPSLGAIL DGPSPEPGQQ LGQGLDDSGN LLSPAPMDWD MLMEPPFLFT AVPPSGELAP PAVPPQAPRC HVVIRGLCGE QP | |
Greater than 90% as determined by SDS-PAGE. |
90113 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |