missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 212 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 212, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_036388.2 |
---|---|
Gene ID (Entrez) | 7988 |
Name | Zinc finger protein 212 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016670
|
enQuireBio™
QP6905-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036670
|
enQuireBio™
QP6905-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026670
|
enQuireBio™
QP6905-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046670
|
enQuireBio™
QP6905-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056670
|
enQuireBio™
QP6905-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066670
|
enQuireBio™
QP6905-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036388.2 | |
Zinc finger protein 212 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7988 | |
Research Use Only | |
ZNF212 | |
Recombinant Protein | |
E. coli | |
MAESAPARHR RKRRSTPLTS STLPSQATEK SSYFQTTEIS LWTVVAAIQA VEKKMESQAA RLQSLEGRTG TAEKKLADCE KMAVEFGNQL EGKWAVLGTL LQEYGLLQRR LENVENLLRN RNFWILRLPP GSKGEAPKVS RSLENDGVCF TEQEWENLED WQKELYRNVM ESNYETLVSL KVLGQTEGEA ELGTEMLGDL EEEGPGGAHP AGGVMIKQEL QYTQEGPADL PGEFSCIAEE QAFLSPEQTE LWGGQGSSVL LETGPGDSTL EEPVGSRVPS SSRTVGCPKQ KSHRQVQLDQ ECGQGLKLKK DTSRPYECSE CEITFRYKQQ LATHLRSHSG WGSCTPEEPE ESLRPRPRLK PQTKKAKLHQ CDVCLRSFSC KVSLVTHQRC HLQEGPSAGQ HVQERFSPNS LVALPGHIPW RKSRSSLICG YCGKSFSHPS DLVRHQRIHT GERPYSCTEC EKSFVQKQHL LQHQKIHQRE RGGLALEPGR PNGLL | |
Greater than 90% as determined by SDS-PAGE. |