missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H1N1 (Puerto Rico / 8 / 1934) NucleoProtein

A DNA sequence encoding the Influenza A virus (strain A/Puerto Rico/8/1934 H1N1) Influenza A H1N1 (strain A/Puerto Rico/8/1934) Nucleoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7280-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077980

  • 12410.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_040982.1
Influenza A H1N1 (Puerto Rico / 8 / 1934) NucleoProtein
Research Use Only
NP
Recombinant Protein
Yeast
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIGGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYRRV NGKWMRELIL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDATYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELVRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQKAMMD QVRESRNPGN AEFEDLTFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEREGYSLVG IDPFRLLQNS QVYSLIRPNE NPAHKSQLVW MACHSAAFED LRVLSFIKGT KVLPRGKLST RGVQIASNEN METMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DRTTIMAAFN GNTEGRTSDM RTEIIRMMES ARPEDVSFQG RGVFELSDEK AASPIVPSFD MSNEGSYFFG DNAEEYDN
Greater than 80% as determined by SDS-PAGE.
956531
100 μg
Not determined.
Full Length
Influenza
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers