missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant M. tuberculosis Steroid C26-monooxygenase Protein

A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Steroid C26-monooxygenase, was expressed in the hosts and tags indicated.

3490.00 SEK - 31480.00 SEK

Specifications

Accession Number NP_218062.1
Gene ID (Entrez) 887782
Name M. tuberculosis Steroid C26-monooxygenase Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16047870
View Documents Promotion Details
enQuireBio™
QP7253-ye-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16087870
View Documents Promotion Details
enQuireBio™
QP7253-ye-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16037870
View Documents Promotion Details
enQuireBio™
QP7253-ye-100ug
100 μg
12935.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16067870
View Documents Promotion Details
enQuireBio™
QP7253-ye-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16077870
View Documents Promotion Details
enQuireBio™
QP7253-ye-500ug
500 μg
31245.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16057870
View Documents Promotion Details
enQuireBio™
QP7253-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_218062.1
M. tuberculosis Steroid C26-monooxygenase Protein
Not determined.
Full Length
M. tuberculosis
His
Tris-based buffer, 50% glycerol
887782
Research Use Only
cyp125
Recombinant Protein
Yeast
MSWNHQSVEI AVRRTTVPSP NLPPGFDFTD PAIYAERLPV AEFAELRSAA PIWWNGQDPG KGGGFHDGGF WAITKLNDVK EISRHSDVFS SYENGVIPRF KNDIAREDIE VQRFVMLNMD APHHTRLRKI ISRGFTPRAV GRLHDELQER AQKIAAEAAA AGSGDFVEQV SCELPLQAIA GLLGVPQEDR GKLFHWSNEM TGNEDPEYAH IDPKASSAEL IGYAMKMAEE KAKNPADDIV TQLIQADIDG EKLSDDEFGF FVVMLAVAGN ETTRNSITQG MMAFAEHPDQ WELYKKVRPE TAADEIVRWA TPVTAFQRTA LRDYELSGVQ IKKGQRVVMF YRSANFDEEV FQDPFTFNIL RNPNPHVGFG GTGAHYCIGA NLARMTINLI FNAVADHMPD LKPISAPERL RSGWLNGIKH WQVDYTGRCP VAH
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications