missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Acid ceramidase Protein

A DNA sequence encoding the Mus musculus (Mouse) Acid ceramidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8262-ec-1mg

 View more versions of this product

Product Code. 16023621

  • 40175.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_062708.1
Acid ceramidase Protein
Research Use Only
ASAH1
Recombinant Protein
E. coli
QAQDVPPWTE DCRKSTYPPS GPTYRGPVPW HTINLDLPPY KRWHELLAQK APALRILVNS ITSLVNTFVP SGKLMKMVDQ KLPGMIGSLP DPFGEEMRGI ADVTGIPLGE IISFNIFYEL FTM
Greater than 80% as determined by SDS-PAGE.
11886
1 mg
Not determined.
Partial (see sequence information for more details).
Mouse
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers