missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Bovine Enterokinase / PRSS7 Protein
A DNA sequence encoding the Mus musculus (Mouse) Bovine Enterokinase / PRSS7, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_032967.1 |
---|---|
Gene ID (Entrez) | 19146 |
Name | Enterokinase / PRSS7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024030
|
enQuireBio™
QP6539-ec-10ug |
10 μg | N/A | ||||||
16064030
|
enQuireBio™
QP6539-ec-50ug |
50 μg | N/A | ||||||
16014030
|
enQuireBio™
QP6539-ec-100ug |
100 μg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16044030
|
enQuireBio™
QP6539-ec-200ug |
200 μg | N/A | ||||||
16054030
|
enQuireBio™
QP6539-ec-500ug |
500 μg | N/A | ||||||
16034030
|
enQuireBio™
QP6539-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_032967.1 | |
Enterokinase / PRSS7 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
19146 | |
Research Use Only | |
Tmprss15 | |
Recombinant Protein | |
E. coli | |
IVGGSDAQAG AWPWVVALYH RDRSTDRLLC GASLVSSDWL VSAAHCVYRR NLDPTRWTAV LGLHMQSNLT SPQVVRRVVD QIVINPHYDR RRKVNDIAMM HLEFKVNYTD YIQPICLPEE NQIFIPGRTC SIAGWGYDKI NAGSTVDVLK EADVPLISNE KCQQQLPEYN ITESMICAGY EEGGIDSCQG DSGGPLMCQE NNRWFLVGVT SFGVQCALPN HPGVYVRVSQ FIEWIHSFLH | |
Greater than 80.8% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title