Learn More
R&D Systems™ Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 24-119) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 528-MP-025/CF
This item is not returnable.
View return policy
Description
The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 24-119) is derived from E. coli. The Recombinant Mouse CCL24/Eotaxin-2/MPIF-2 (aa 24-119) has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
10.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
56221 | |
CCL24/Eotaxin-2/MPIF-2 aa 24-119 | |
E. coli-derived mouse CCL24/Eotaxin-2/MPIF-2 protein Ile24-Val119 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |