Learn More
R&D Systems™ Recombinant Mouse Fc gamma RIII/CD16 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 1960-FC-050
This item is not returnable.
View return policy
Description
The Recombinant Mouse Fc gamma RIII/CD16 Protein is derived from NS0. The Recombinant Mouse Fc gamma RIII/CD16 Protein has been validated for the following applications: Binding Activity.
Specifications
P08508.1 | |
14131 | |
Fc gamma RIII (CD16) | |
Mouse myeloma cell line,NS0-derived mouse Fc gamma RIII (CD16) protein Ala31-Thr215,with a C-terminal 10-His tag | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Supplied as a 0.2μm filtered solution in PBS. | |
30-45 kDa, reducing conditions | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. | |
Recombinant |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
R&D Systems™ Recombinant Mouse Fc gamma RIII/CD16 Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction