missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL-1 beta / IL1B Protein

A DNA sequence encoding the Mus musculus (Mouse) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8893-ec-500ug

 View more versions of this product

Product Code. 16037031

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_032387.1
IL-1 beta / IL1B Protein
Research Use Only
IL1B
Recombinant Protein
E. coli
PIRQLHYRLR DEQQKSLVLS DPYELKALHL NGQNINQQVI FSMSFVQGEP SNDKIPVALG LKGKNLYLSC VMKDGTPTLQ LESVDPKQYP KKKMEKRFVF NKIEVKSKVE FESAEFPNWY ISTSQAEHKP VFLGNNSGQD IIDFTMESVS
Greater than 90% as determined by SDS-PAGE.
16176
500 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers