missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse LY-96 / ESOP-1 Protein

A DNA sequence encoding the Mus musculus (Mouse) LY-96 / ESOP-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6327-ec-200ug

 View more versions of this product

Product Code. 16052500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001153183.1
LY-96 / ESOP-1 Protein
Research Use Only
LY96
Recombinant Protein
E. coli
EKQQWFCNSS DAIISYSYCD HLKFPISISS EPCIRLRGTN GFVHVEFIPR GNLKYLYFNL FISVNSIELP KRKEVLCHGH DDDYSFCRAL KGETVNTSIP FSFEGILFPK GHYRCVAEAI AGDTEEKLFC LNFTIIHRRD VN
Greater than 90% as determined by SDS-PAGE.
17087
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers