missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse OTC Protein
A DNA sequence encoding the Mus musculus (Mouse) OTC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6457-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_032795.1 | |
OTC Protein | |
Research Use Only | |
OTC | |
Recombinant Protein | |
E. coli | |
SQVQLKGRDL LTLKNFTGEE IQYMLWLSAD LKFRIKQKGE YLPLLQGKSL GMIFEKRSTR TRLSTETGFA LLGGHPSFLT TQDIHLGVNE SLTDTARVLS SMTDAVLARV YKQSDLDTLA KEASIPIVNG LSDLYHPIQI LADYLTLQEH YGSLKGLTLS WIGDGNNILH SIMMSAAKFG MHLQAATPKG YEPDPNIVKL AEQYAKENGT KLSMTNDPLE AARGGNVLIT DTWISMGQED EKKKRLQAFQ GYQVTMKTAK VAASDWTFLH CLPRKPEEVD DEVFYSPRSL VFPEAENRKW TIMAVMVSLL TDYSPVLQKP KF | |
Greater than 90% as determined by SDS-PAGE. |
18416 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |