missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse S100A8 / CAGA Protein
A DNA sequence encoding the Mus musculus (Mouse) S100A8 / CAGA, was expressed in the hosts and tags indicated.
2430.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_038678.1 |
---|---|
Gene ID (Entrez) | 20201 |
Name | S100A8 / CAGA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094740
|
enQuireBio™
QP6641-ye-10ug |
10 μg | N/A | ||||||
16034750
|
enQuireBio™
QP6641-ye-50ug |
50 μg | N/A | ||||||
16084740
|
enQuireBio™
QP6641-ye-100ug |
100 μg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16014750
|
enQuireBio™
QP6641-ye-200ug |
200 μg | N/A | ||||||
16024750
|
enQuireBio™
QP6641-ye-500ug |
500 μg |
25885.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16004750
|
enQuireBio™
QP6641-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_038678.1 | |
S100A8 / CAGA Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20201 | |
Research Use Only | |
S100A8 | |
Recombinant Protein | |
Yeast | |
PSELEKALSN LIDVYHNYSN IQGNHHALYK NDFKKMVTTE CPQFVQNINI ENLFRELDIN SDNAINFEEF LAMVIKVGVA SHKDSHKE | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title