missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Survivin / BIRC5 / API4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5720-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001012273.1 | |
Survivin / BIRC5 / API4 Protein | |
Research Use Only | |
BIRC5 | |
Recombinant Protein | |
E. coli | |
MGAPALPQIW QLYLKNYRIA TFKNWPFLED CACTPERMAE AGFIHCPTEN EPDLAQCFFC FKELEGWEPD DNPIEEHRKH SPGCAFLTVK KQMEELTVSE FLKLDRQRAK NKIAKETNNK QKEFEETAKT TRQSIEQLAA | |
Greater than 90% as determined by SDS-PAGE. |
11799 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse Survivin / BIRC5 / API4 Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction