missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse TNF-alpha Protein

A DNA sequence encoding the Mus musculus (Mouse) TNF-alpha, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6811-ec-200ug

 View more versions of this product

Product Code. 16046010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001265530.1
TNF-alpha Protein
Research Use Only
TNF
Recombinant Protein
E. coli
GPQRDEKFPN GLPLISSMAQ TLTLRSSSQN SSDKPVAHVV ANHQVEEQLE WLSQRANALL ANGMDLKDNQ LVVPADGLYL VYSQVLFKGQ GCPDYVLLTH TVSRFAISYQ EKVNLLSAVK SPCPKDTPEG AELKPWYEPI YLGGVFQLEK GDQLSAEVNL PKYLDFAESG QVYFGVIAL
Greater than 90% as determined by SDS-PAGE.
21926
200 μg
Not determined.
Extracellular Domain
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers