missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma High affinity transport system protein p37 Protein
A DNA sequence encoding the Mycoplasma hyorhinis High affinity transport system protein p37, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7079-200ug
This item is not returnable.
View return policy
Specifications
Mycoplasma High affinity transport system protein p37 Protein | |
Research Use Only | |
p37 | |
Recombinant Protein | |
E. coli | |
CSNTGVVKQE DVSVSQGQWD KSITFGVSEA WLNKKKGGEK VNKEVINTFL ENFKKEFNKL KNANDKTKNF DDVDFKVTPI QDFTVLLNNL STDNPELDFG INASGKLVEF LKNNPGIITP ALETTTNSFV FDKEKDKFYV DGTDSDPLVK IAKEINKIFV ETPYASWTDE NHKWNGNVYQ SVYDPTVQAN FYRGMIWIKG NDETLAKIKK AWNDKDWNTF RNFGILHGKD NSFSKFKLEE TILKNHFQNK FTTLNEDRSA HPNAYKQKSA DTLGTLDDFH IAFSEEGSFA WTHNKSATKP FETKANEKME ALIVTNPIPY DVGVFRKSVN QLEQNLIVQT FINLAKNKQD TYGPLLGYNG YKKIDNFQKE IVEVYEKAIK | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |