missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Transcobalamin-1 Protein

A DNA sequence encoding the Sus scrofa (Pig) Transcobalamin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9423-ye-500ug

 View more versions of this product

Product Code. 16018281

  • 31245.00 SEK / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Transcobalamin-1 Protein
Research Use Only
TCN1
Recombinant Protein
Yeast
CVVSEKDYSH LRLLISAMDN LEQIRGIYGA SILLSQRLAG IQNPSLEEEL SQRIQDDMNR RDMSNLTSGQ LALIILAFGA CKTPDVRFIH DHHLVEKLGE KFKEEIKNME IHNSNPLTNY YQLSFDVLTL CLFRGNYSIS NVTHYFNPEN KNFNLSGHFS VDTGAVAVLA LTCVKRSISN GKIKAAIKDS DTIQKYIESL VHKIQSEKMV VSLETRIAQE KLCRLSLSHQ TITKMNQIAK KLWTRCLTHS QGVFRLPIAA AQILPALLGK TYLDVTKLLL VPKVQVNITD EPVPVVPTLS PENISVIYCV KINEISNCIN ITVFLDVMKA AQEKNSTIYG FTMTETPWGP YITSVQGIWA NNNERTYWEH SEQQQITKPR SMGIMLSKME SI
Greater than 80% as determined by SDS-PAGE.
500 μg
Not determined.
Full Length of Mature Protein
Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers