missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BD 1 Protein
A cDNA sequence encoding the BD 1 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Gene ID (Entrez) | 83687 |
---|---|
Name | BD 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Defb1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934158
|
enQuireBio™
QP10531-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924158
|
enQuireBio™
QP10531-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914158
|
enQuireBio™
QP10531-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
83687 | |
Research Use Only | |
Defb1 | |
Recombinant Protein | |
E. coli | |
DQYRCLQNGG FCLRSSCPSH TKLQGTCKPD KPNCCRS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
BD 1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its ability to chemoattract CD34+ dendritic cells using a concentration range of 0.1-1.0 ug/ml. | |
Rat | |
Untagged | |
BD-1 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |