missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Collagen alpha-1(I) chain Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Collagen alpha-1(I) chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9077-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017581

  • 8695.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_445756.1
Collagen alpha-1(I) chain Protein
Research Use Only
Col1a1
Recombinant Protein
Yeast
QRGERGFPGL PGPSGEPGKQ GPSGASGERG PPGPMGPPGL AGPPGESGRE GSPGAEGSPG RDGAPGAKGD RGETGPAGPP GAPGAPGAPG PVGPAGKNGD RGETGPAGPA GPIGPAGARG PAGPQGPRGD KGETGEQGDR GIKGHRGFSG LQGPPGSPGS PGEQGPSGAS GPAGPRGPPG SAGSPGKDGL NGLPGPIGPP GPRGRTGDSG PAGPPGPPGP PGPPGPPSGG YDFSFLPQPP QEKSQDGGRY YRA
Greater than 90% as determined by SDS-PAGE.
29393
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers