Learn More
R&D Systems™ Recombinant Rat FGF-BP Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand:  R&D Systems™ 1413-FB-025/CF
 View more versions of this product
Product Code. 18107060
  • 3095.00 SEK / 25µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Recombinant Rat FGF-BP Protein is derived from NS0. The Recombinant Rat FGF-BP Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS.
26 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
64535
FGF-BP
Mouse myeloma cell line,NS0-derived rat FGF-BP protein Glu21-Cys238,with a C-terminal Asp and 10-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.