missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Hemopexin / HPX Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Hemopexin / HPX, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_445770.1 |
---|---|
Gene ID (Entrez) | 58917 |
Name | Hemopexin / HPX Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16021370
|
enQuireBio™
QP6180-ec-10ug |
10 μg | N/A | ||||||
16061370
|
enQuireBio™
QP6180-ec-50ug |
50 μg | N/A | ||||||
16011370
|
enQuireBio™
QP6180-ec-100ug |
100 μg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16041370
|
enQuireBio™
QP6180-ec-200ug |
200 μg | N/A | ||||||
16051370
|
enQuireBio™
QP6180-ec-500ug |
500 μg | N/A | ||||||
16031370
|
enQuireBio™
QP6180-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_445770.1 | |
Hemopexin / HPX Protein | |
Not determined. | |
Full Length | |
Rat | |
Tris-based buffer, 50% glycerol |
58917 | |
Research Use Only | |
HPX | |
Recombinant Protein | |
NPLPAAHETV AKGENGTKPD SDVIEHCSDA WSFDATTMDH NGTMLFFKGE FVWRGHSGIR ELISERWKNP VTSVDAAFRG PDSVFLIKED KVWVYPPEKK ENGYPKLFQE EFPGIPYPPD AAVECHRGEC QSEGVLFFQG NRKWFWDFAT RTQKERSWPA VGNCTAALRW LERYYCFQGN KFLRFNPVTG EVPPRYPLDA RDYFISCPGR GHGKLRNGTA HGNSTHPMHS RCNADPGLSA LLSDHRGATY AFSGSHYWRL DSSRDGWHSW PIAHHWPQGP SAVDAAFSWD EKVYLIQGTQ VYVFLTKGGN NLVSGYPKRL EKELGSPPGI SLDTIDAAFS CPGSSKLYVT SGRRLWWLDL KSGAQATWAE LSWPHEKVDG ALCLEKSLGP YSCSSNGPNL FFIHGPNLYC YSSIDKLNAA KSLPQPQKVN SILGCSQ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title