missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Monoglyceride lipase Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Monoglyceride lipase, was expressed in the hosts and tags indicated.
Specifications
Accession Number | NP_612511.1 |
---|---|
Gene ID (Entrez) | 29254 |
Name | Monoglyceride lipase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087971
|
enQuireBio™
QP9272-ye-10ug |
10 μg |
N/A
|
N/A | |||||
16027981
|
enQuireBio™
QP9272-ye-50ug |
50 μg |
N/A
|
N/A | |||||
16077971
|
enQuireBio™
QP9272-ye-100ug |
100 μg |
N/A
|
N/A | |||||
16007981
|
enQuireBio™
QP9272-ye-200ug |
200 μg |
N/A
|
N/A | |||||
16017981
|
enQuireBio™
QP9272-ye-500ug |
500 μg |
N/A
|
N/A | |||||
16097971
|
enQuireBio™
QP9272-ye-1mg |
1 mg |
N/A
|
N/A | |||||
Specifications
NP_612511.1 | |
Monoglyceride lipase Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
29254 | |
Research Use Only | |
Mgll | |
Recombinant Protein | |
Yeast | |
MPEASSPRRT PQNVPYQDLP HLVNADGQYL FCRYWKPSGT PKALIFVSHG AGEHCGRYDE LAQMLKRLDM LVFAHDHVGH GQSEGERMVV SDFQVFVRDL LQHVNTVQKD YPEVPVFLLG HSMGGAISIL AAAERPTHFS GMILISPLIL ANPESASTLK VLAAKLLNFV LPNISLGRID SSVLSRNKSE VDLYNSDPLI CHAGVKVCFG IQLLNAVSRV ERAMPRLTLP FLLLQGSADR LCDSKGAYLL MESSPSQDKT LKMYEGAYHV LHKELPEVTN SVLHEINTWV SHRIAVAGAR CLP | |
Greater than 90% as determined by SDS-PAGE. |