missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Rat PBEF/Visfatin/NAMPT FLAG Protein
Recombinant Protein
2590.00 SEK - 5880.00 SEK
Specifications
Accession Number | Q80Z29 |
---|---|
Concentration | 0.35 mg/ml |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. 0.2um-filtered solution in PBS, pH 7.2 |
Gene ID (Entrez) | 10135 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18323725
|
Novus Biologicals™
NBP3-18142-50UG |
50 μg |
5880.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18389706
|
Novus Biologicals™
NBP3-18142-10UG |
10 μg |
2590.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Q80Z29 | |
SDS-PAGE | |
10135 | |
Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles. | |
EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity | |
PBEF/Visfatin/NAMPT |
0.35 mg/ml | |
Liquid. 0.2um-filtered solution in PBS, pH 7.2 | |
M.W. Theoretical: 60 kDa | |
<0.1 EU/μg purified protein (LAL test; Lonza). | |
NAMPT | |
>90% SDS-PAGE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title