missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat SerpinE1 / PAI-1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) SerpinE1 / PAI-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6673-ec-100ug

 View more versions of this product

Product Code. 16005000

  • 9055.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_036752.1
SerpinE1 / PAI-1 Protein
Research Use Only
Serpine1
Recombinant Protein
E. coli
SPLPESHTAQ QATNFGVKVF QHVVQASKDR NVVFSPYGVS SVLAMLQLTT AGKTRQQIQD AMGFNISERG TAPALRKLSK ELMGSWNKNE ISTADAIFVQ RDLELVQGFM PHFFKLFRTT VKQVDFSEVE RARFIINDWV ERHTKGMISD LLAKGAVNEL TRLVLVNALY FNGQWKTPFL EASTHQRLFH KSDGSTISVP MMAQNNKFNY TEFTTPDGHE YDILELPYHG ETLSMFIAAP FEKDVPLSAI TNILDAELIR QWKSNMTRLP RLLILPKFSL ETEVDLRGPL EKLGMTDIFS STQADFTSLS DQEQLSVAQA LQKVKIEVNE SGTVASSSTA ILVSARMAPT EMVLDRSFLF VVRHNPTETI LFMGQLMEP
Greater than 90% as determined by SDS-PAGE.
24617
100 μg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers