missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Zinc transporter ZIP13 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Zinc transporter ZIP13, was expressed in the hosts and tags indicated.
2430.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_001034285.1 |
---|---|
Gene ID (Entrez) | 295928 |
Name | Zinc transporter ZIP13 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009950
|
enQuireBio™
QP7638-ec-10ug |
10 μg | N/A | ||||||
16049950
|
enQuireBio™
QP7638-ec-50ug |
50 μg | N/A | ||||||
16099940
|
enQuireBio™
QP7638-ec-100ug |
100 μg |
9055.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16029950
|
enQuireBio™
QP7638-ec-200ug |
200 μg | N/A | ||||||
16039950
|
enQuireBio™
QP7638-ec-500ug |
500 μg | N/A | ||||||
16019950
|
enQuireBio™
QP7638-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001034285.1 | |
Zinc transporter ZIP13 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-GST | |
Tris-based buffer, 50% glycerol |
295928 | |
Research Use Only | |
Slc39a13 | |
Recombinant Protein | |
E. coli | |
WAYTCNISPG VEGQSLQRQQ QLGLWVIAGF LTFLALEKMF LNCKEEDPSQ APSKDPTAAA LNGGHCLAQP AAEPGLRAVV RNLKVSGYLN LLANTIDNFT HGLA | |
Greater than 90% as determined by SDS-PAGE. |