missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus E.coli Enoyl-ACP Reductase / FABI Protein
A DNA sequence encoding the Staphylococcus aureus (strain MRSA252) E.coli Enoyl-ACP Reductase / FABI, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7753-ec-50ug
This item is not returnable.
View return policy
Specifications
WP_000933195.1 | |
S. aureus E.coli Enoyl-ACP Reductase / FABI Protein | |
Research Use Only | |
fabI | |
Recombinant Protein | |
E. coli | |
MLNLENKTYV IMGIANKRSI AFGVAKVLDQ LGAKLVFTYR KERSRKELEK LLEQLNQPEA HLYQIDVQSD EEVINGFEQI GKDVGNIDGV YHSIAFANME DLRGRFSETS REGFLLAQDI SSYSLTIVAH EAKKLMPEGG SIVATTYLGG EFAVQNYNVM GVAKASLEAN VKYLALDLGP DNIRVNAISA GPIRTLSAKG VGGFNTILKE IEERAPLKRN VDQVEVGKTA AYLLSDLSSG VTGENIHVDS GFHAIK | |
Greater than 90% as determined by SDS-PAGE. |
31213761 | |
50 μg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |