missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus esxA Protein

A DNA sequence encoding the Staphylococcus aureus (strain MSSA476) esxA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7776-ec-500ug

 View more versions of this product

Product Code. 16060721

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

WP_001240826.1
S. aureus esxA Protein
Research Use Only
esxA
Recombinant Protein
E. coli
MAMIKMSPEE IRAKSQSYGQ GSDQIRQILS DLTRAQGEIA ANWEGQAFSR FEEQFQQLSP KVEKFAQLLE EIKQQLNSTA DAVQEQDQQL SNNFGLQ
Greater than 90% as determined by SDS-PAGE.
31213083
500 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers