missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Alanine / arginine aminopeptidase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alanine/arginine aminopeptidase, was expressed in the hosts and tags indicated.
5195.00 SEK - 44210.00 SEK
Specifications
Accession Number | NP_011913.1 |
---|---|
Gene ID (Entrez) | 856443 |
Name | S. cerevisiae Alanine / arginine aminopeptidase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087450
|
enQuireBio™
QP7127-10ug |
10 μg |
5195.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16027460
|
enQuireBio™
QP7127-50ug |
50 μg |
7415.00 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16077450
|
enQuireBio™
QP7127-100ug |
100 μg |
12630.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16007460
|
enQuireBio™
QP7127-200ug |
200 μg |
20825.00 SEK
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16017460
|
enQuireBio™
QP7127-500ug |
500 μg |
27820.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16097450
|
enQuireBio™
QP7127-1mg |
1 mg |
44210.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_011913.1 | |
S. cerevisiae Alanine / arginine aminopeptidase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
856443 | |
Research Use Only | |
AAP1 | |
Recombinant Protein | |
E. coli | |
MSREVLPNNV TPLHYDITLE PNFRAFTFEG SLKIDLQIND HSINSVQINY LEIDFHSARI EGVNAIEVNK NENQQKATLV FPNGTFENLG PSAKLEIIFS GILNDQMAGF YRAKYTDKVT GETKYMATTQ MEATDARRAF PCFDEPNLKA TFAVTLVSES FLTHLSNMDV RNETIKEGKK YTTFNTTPKM STYLVAFIVA DLRYVESNNF RIPVRVYSTP GDEKFGQFAA NLAARTLRFF EDTFNIEYPL PKMDMVAVHE FSAGAMENWG LVTYRVIDLL LDIENSSLDR IQRVAEVIQH ELAHQWFGNL VTMDWWEGLW LNEGFATWMS WYSCNKFQPE WKVWEQYVTD NLQRALNLDS LRSSHPIEVP VNNADEINQI FDAISYSKG | |
Greater than 80% as determined by SDS-PAGE. |