missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Dengue Virus 4 VLP Protein
15435.00 SEK - 54845.00 SEK
Specifications
For Use With (Application) | SDS-PAGE |
---|---|
Formulation | Dulbecco's PBS pH7.4, 30% sucrose |
Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Purity or Quality Grade | >95% pure by SDS-PAGE |
Protein | Dengue Virus 4 VLP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18395192
|
Novus Biologicals
NBP3-14810-0.1MG |
0.1 mg |
15435.00 SEK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18324095
|
Novus Biologicals
NBP3-14810-0.25MG |
0.25 mg |
30250.00 SEK
0.25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18359624
|
Novus Biologicals
NBP3-14810-0.5MG |
0.5 mg |
54845.00 SEK
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SDS-PAGE | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
Dengue Virus 4 VLP |
Dulbecco's PBS pH7.4, 30% sucrose | |
>95% pure by SDS-PAGE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title