Learn More
Redox Standard 200mV, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Packaging | 500mL |
Quantity | 500 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
RS20024B1 | Certificate of Analysis | 11/03/2025 | RS200 |
RS20023D1 | Certificate of Analysis | 12/12/2023 | RS200 |
RS20021K1 | Certificate of Analysis | 01/03/2022 | RS200 |
RS20020L1 | Certificate of Analysis | 24/08/2021 | RS200 |
RS20022A1 | Certificate of Compliance | 05/08/2022 | RS200 |
Product Content Correction
Product Title
Redox Standard 200mV, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction