Learn More
Redox Standard 358mV, Reagecon™ Product Code.: 13138736
Redox Standard 358mV, Reagecon™
Product Code. 13138736 Shop All Reagecon Products
500 mL
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
Product Code. 13138736
Brand: Reagecon RS358
Specifications
Packaging 500mL
Quantity 500 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberRS35824K1 Certificate TypeCertificate of Analysis Date15/03/2025 Catalog NumberRS358
Lot NumberRS35823H1 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog NumberRS358
Lot NumberRS35822D1 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog NumberRS358
Product Content Correction
Product Title
Redox Standard 358mV, Reagecon™ >