missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REEP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1620.00 SEK - 4750.00 SEK
Specifications
Antigen | REEP1 |
---|---|
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321435
|
Bio-Techne
NBP3-14647-50UL |
50 μg |
1620.00 SEK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30032503
|
Novus Biologicals
NBP3-14647-100ul |
100 μL |
4750.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
REEP1 Polyclonal antibody specifically detects REEP1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
REEP1 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
C2orf23, chromosome 2 open reading frame 23, receptor accessory protein 1, receptor expression-enhancing protein 1, SPG31FLJ13110 | |
Produced in rabbits immunized with E. coli-derived Human REEP1 fragment | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuroscience | |
PBS, pH 7.0 with 0.03% Proclin 300 | |
65055 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title