Learn More
Thermo Scientific™ Remel™ Isolator Wrap™ Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
1066.50 SEK
Specifications
Description Sabouraud Dextrose Agar
Format Foil Barrier Wrap + Bag
Product Type Agar
View More Specs
Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17772608
View Documents Promotion Details
Thermo Scientific™
R111825
10/Pk.
1066.50 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Remel Isolator Wrap Sterile Sabauroud Dextrose Agar Contact Plate w/Lecithin, Polysorbate 80 is used for the cultivation of yeasts and fungi.
  • Foil Barrier Wrap — impermeable to isolator decontamination process
  • GripFit™ Plates — easy to handle with one hand without lid separating from bottom of plate
  • Irradiated — Sterility Assurance Level 10-6
  • Shrink-wraped with easy open tear strip — reduces breakage and moisture loss
  • Bottom labeling — information stays with sample
  • Extra sterile bag — easy to transport plates to lab without introducing contamination
  • Six month shelf life — less ordering and fewer lot numbers to QC
Specifications
Specifications
Sabouraud Dextrose Agar
Foil Barrier Wrap + Bag
Agar
Documents
Product Certifications
Not intended for IVD use.