missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar with Lecithin and Polysorbate 80

A sterile environmental monitoring 100 mm TSA plate for testing microbial limits in maintaining cleanrooms and manufacturing facilities

Brand:  Thermo Scientific™ R111892

Additional Details : Weight : 0.35000kg

Product Code. 17465539

  • 1046.21 SEK / Pack of 10
Estimated Shipment: 03-10-2024
to see stock.
Add to basket

Description

Description

  • Sterile plated culture media
  • Unique 100mm plate
  • Double bagged typek pouch
  • Convenient packaging for transport
  • Confidence of no contamination in cleanroom
  • Media that grows microorganisms which are compromised to confirm cleanroom standards
Specifications

Specifications

100 mm Plate
Agar
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers